SY3H3D68115D Synthonix Corporation

Methyl 2-(2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate

  • Cas Number : 1415960-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58820.html

Safety Information

MSDS https://synthonix.com/58820.html
Availability details for
Quantity
  • Availability
  • Add to Cart