SY3H3D68115E Synthonix Corporation

(S)-3-Methyl-1-(2-(piperidin-1-yl)phenyl)butan-1-amine

  • Cas Number : 147769-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58840.html

Safety Information

MSDS https://synthonix.com/58840.html
Availability details for
Quantity
  • Availability
  • Add to Cart