SY3H3D681161 Synthonix Corporation

Methyl 1,4-Bis(Boc)-2-piperazinecarboxylate

  • MDL number No MDL found
  • Cas Number : 171504-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58852.html

Safety Information

MSDS https://synthonix.com/58852.html
Availability details for
Quantity
  • Availability
  • Add to Cart