SY3H3D681162 Synthonix Corporation

2-Methyldibenzo[f,h]quinoxaline

  • Cas Number : 536753-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58894.html

Safety Information

MSDS https://synthonix.com/58894.html
Availability details for
Quantity
  • Availability
  • Add to Cart