SY3H3D681163 Synthonix Corporation

2-(Methoxy(4-phenoxyphenyl)methylene)malononitrile

  • MDL number No MDL found
  • Cas Number : 330792-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58895.html

Safety Information

MSDS https://synthonix.com/58895.html
Availability details for
Quantity
  • Availability
  • Add to Cart