SY3H3D681164 Synthonix Corporation

Methyl 4-hydroxy-7-phenoxyisoquinoline-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 1455091-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58901.html

Safety Information

MSDS https://synthonix.com/58901.html
Availability details for
Quantity
  • Availability
  • Add to Cart