SY3H3D681169 Synthonix Corporation

((2S,5R)-5-(5-Methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-2,5-dihydrofuran-2-yl)methyl benzoate

  • MDL number No MDL found
  • Cas Number : 122567-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58929.html

Safety Information

MSDS https://synthonix.com/58929.html
Availability details for
Quantity
  • Availability
  • Add to Cart