SY3H3D68116B Synthonix Corporation

6-Methoxy-2-phenyl-3,4-dihydronaphthalen-1(2H)-one

  • Cas Number : 1769-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58939.html

Safety Information

MSDS https://synthonix.com/58939.html
Availability details for
Quantity
  • Availability
  • Add to Cart