SY3H3D68116D Synthonix Corporation

4-Methoxyphenyl 4-propylbenzoate

  • MDL number No MDL found
  • Cas Number : 50649-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58967.html

Safety Information

MSDS https://synthonix.com/58967.html
Availability details for
Quantity
  • Availability
  • Add to Cart