SY3H3D68116E Synthonix Corporation

Methyl 3-(4-(benzyloxy)phenyl)propanoate

  • MDL number No MDL found
  • Cas Number : 24807-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58968.html

Safety Information

MSDS https://synthonix.com/58968.html
Availability details for
Quantity
  • Availability
  • Add to Cart