SY3H3D681170 Synthonix Corporation

Methyl 3-methoxy-4-((4-methoxybenzyl)oxy)benzoate

  • MDL number No MDL found
  • Cas Number : 1354549-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58970.html

Safety Information

MSDS https://synthonix.com/58970.html
Availability details for
Quantity
  • Availability
  • Add to Cart