SY3H3D681171 Synthonix Corporation

2-(4-Methyl-2-phenylpiperazin-1-yl)nicotinic acid

  • MDL number No MDL found
  • Cas Number : 61338-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58979.html

Safety Information

MSDS https://synthonix.com/58979.html
Availability details for
Quantity
  • Availability
  • Add to Cart