SY3H3D681172 Synthonix Corporation

N-Methyl-2-(4-nitrophenoxy)-N-[2-(4-nitrophenyl)ethyl]ethanamine

  • Cas Number : 115287-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58981.html

Safety Information

MSDS https://synthonix.com/58981.html
Availability details for
Quantity
  • Availability
  • Add to Cart