SY3H3D681176 Synthonix Corporation

2-(2-Methoxyphenoxy)-1-(4-methoxyphenyl)propane-1,3-diol

  • MDL number No MDL found
  • Cas Number : 92409-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59006.html

Safety Information

MSDS https://synthonix.com/59006.html
Availability details for
Quantity
  • Availability
  • Add to Cart