SY3H3D681177 Synthonix Corporation

4-(4-(4-Methoxyphenyl)piperazin-1-yl)aniline

  • Cas Number : 74852-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/59011.html

Safety Information

MSDS https://synthonix.com/59011.html
Availability details for
Quantity
  • Availability
  • Add to Cart