SY3H3D68117A Synthonix Corporation

2-Methyl-4-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)butan-2-ol

  • MDL number No MDL found
  • Cas Number : 1362243-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59089.html

Safety Information

MSDS https://synthonix.com/59089.html
Availability details for
Quantity
  • Availability
  • Add to Cart