SY3H3D68117F Synthonix Corporation

(E)-Methyl 3-(methoxy(phenyl)methylene)-2-oxoindoline-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 1168150-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59182.html

Safety Information

MSDS https://synthonix.com/59182.html
Availability details for
Quantity
  • Availability
  • Add to Cart