SY3H3D681181 Synthonix Corporation

1-Methoxy-4-((4-propylphenyl)ethynyl)benzene

  • Cas Number : 39969-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59212.html

Safety Information

MSDS https://synthonix.com/59212.html
Availability details for
Quantity
  • Availability
  • Add to Cart