SY3H3D681182 Synthonix Corporation

1-(3-(Methylamino)-1-(thiophen-2-yl)propyl)naphthalen-2-ol

  • Cas Number : 1346599-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59226.html

Safety Information

MSDS https://synthonix.com/59226.html
Availability details for
Quantity
  • Availability
  • Add to Cart