SY3H3D681184 Synthonix Corporation

6-(4-Methylpiperazin-1-yl)-4-(o-tolyl)nicotinamide

  • Cas Number : 342417-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59259.html

Safety Information

MSDS https://synthonix.com/59259.html
Availability details for
Quantity
  • Availability
  • Add to Cart