SY3H3D68118A Synthonix Corporation

N-(4-Methoxybenzyl)-1-(naphthalen-1-yl)methanamine

  • MDL number No MDL found
  • Cas Number : 185669-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59399.html

Safety Information

MSDS https://synthonix.com/59399.html
Availability details for
Quantity
  • Availability
  • Add to Cart