SY3H3D68118C Synthonix Corporation

Methyl 1-Benzyl-4-(Boc-amino)piperidine-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 294180-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59444.html

Safety Information

MSDS https://synthonix.com/59444.html
Availability details for
Quantity
  • Availability
  • Add to Cart