SY3H3D68118D Synthonix Corporation

(S)-Methyl 3-((tert-butoxycarbonyl)amino)-3-methyl-2-(((S)-1-phenylethyl)amino)butanoate

  • MDL number No MDL found
  • Cas Number : 1093192-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/59454.html

Safety Information

MSDS https://synthonix.com/59454.html
Availability details for
Quantity
  • Availability
  • Add to Cart