SY3H3D68118F Synthonix Corporation

3-((2S,4S)-4-Mercapto-1-(((4-nitrobenzyl)oxy)carbonyl)pyrrolidine-2-carboxamido)benzoic acid

  • MDL number No MDL found
  • Cas Number : 202467-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59524.html

Safety Information

MSDS https://synthonix.com/59524.html
Availability details for
Quantity
  • Availability
  • Add to Cart