SY3H3D681190 Synthonix Corporation

4-Methoxy-N-(4-methoxyphenyl)-N-phenylaniline

  • MDL number No MDL found
  • Cas Number : 20440-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59529.html

Safety Information

MSDS https://synthonix.com/59529.html
Availability details for
Quantity
  • Availability
  • Add to Cart