SY3H3D68119C Synthonix Corporation

3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid

  • MDL number MFCD00191556
  • Cas Number : 77359-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39646.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39646.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart