SY3H3D6811A2 Synthonix Corporation

3-Nitro-4,5-dihydroisoxazole

  • MDL number No MDL found
  • Cas Number : 1121-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39838.html

Safety Information

MSDS https://synthonix.com/39838.html
Availability details for
Quantity
  • Availability
  • Add to Cart