SY3H3D6811A4 Synthonix Corporation

5-Nitro-1H-imidazole-2-carboxylic acid

  • Cas Number : 351990-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40057.html

Safety Information

MSDS https://synthonix.com/40057.html
Availability details for
Quantity
  • Availability
  • Add to Cart