SY3H3D6811A8 Synthonix Corporation

4-Nitro-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid

  • Cas Number : 1322805-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40492.html

Safety Information

MSDS https://synthonix.com/40492.html
Availability details for
Quantity
  • Availability
  • Add to Cart