SY3H3D6811AA Synthonix Corporation

5-Nitropyridin-3-amine

  • MDL number No MDL found
  • Cas Number : 934-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40963.html

Safety Information

MSDS https://synthonix.com/40963.html
Availability details for
Quantity
  • Availability
  • Add to Cart