SY3H3D6811AC Synthonix Corporation

1-(4-Nitro-1H-pyrazol-5-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 2228374-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40977.html

Safety Information

MSDS https://synthonix.com/40977.html
Availability details for
Quantity
  • Availability
  • Add to Cart