SY3H3D6811AD Synthonix Corporation

5-Nitro-6-methyluracil

  • Cas Number : 16632-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40978.html

Safety Information

MSDS https://synthonix.com/40978.html
Availability details for
Quantity
  • Availability
  • Add to Cart