SY3H3D6811B1 Synthonix Corporation

5-Nitro-2,3-pyridinediamine

  • Cas Number : 3537-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41105.html

Safety Information

MSDS https://synthonix.com/41105.html
Availability details for
Quantity
  • Availability
  • Add to Cart