SY3H3D6811B3 Synthonix Corporation

3-Nitro-1H-pyrazole-1-ethanol

  • Cas Number : 956951-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41154.html

Safety Information

MSDS https://synthonix.com/41154.html
Availability details for
Quantity
  • Availability
  • Add to Cart