SY3H3D6811B4 Synthonix Corporation

4-Nitrobenzene-1,3-diamine sulfate

  • MDL number No MDL found
  • Cas Number : 200295-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41420.html

Safety Information

MSDS https://synthonix.com/41420.html
Availability details for
Quantity
  • Availability
  • Add to Cart