SY3H3D6811B8 Synthonix Corporation

2-Nitropyridine-5-carboxylic acid

  • Cas Number : 33225-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42123.html

Safety Information

MSDS https://synthonix.com/42123.html
Availability details for
Quantity
  • Availability
  • Add to Cart