SY3H3D6811B9 Synthonix Corporation

3-Nitro-6-(trifluoromethyl)pyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 893444-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42169.html

Safety Information

MSDS https://synthonix.com/42169.html
Availability details for
Quantity
  • Availability
  • Add to Cart