SY3H3D6811BA Synthonix Corporation

5-Nitro-3-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD18382741
  • Cas Number : 1121056-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42170.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42170.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart