SY3H3D6811BE Synthonix Corporation

1-(5-Nitrothiophen-2-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 39565-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42610.html

Safety Information

MSDS https://synthonix.com/42610.html
Availability details for
Quantity
  • Availability
  • Add to Cart