SY3H3D6811C5 Synthonix Corporation

5-Nitrobenzene-1,3-diamine

  • MDL number MFCD00025291
  • Cas Number : 5042-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43102.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43102.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart