SY3H3D6811C6 Synthonix Corporation

3-(Nitromethyl)cyclopentanone

  • MDL number No MDL found
  • Cas Number : 81266-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43386.html

Safety Information

MSDS https://synthonix.com/43386.html
Availability details for
Quantity
  • Availability
  • Add to Cart