SY3H3D6811C7 Synthonix Corporation

3-Nitroaniline hydrochloride

  • Cas Number : 33240-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43520.html

Safety Information

MSDS https://synthonix.com/43520.html
Availability details for
Quantity
  • Availability
  • Add to Cart