SY3H3D6811C8 Synthonix Corporation

Nicotinimidamide hydrochloride

  • MDL number MFCD00054326
  • Cas Number : 7356-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43530.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43530.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart