SY3H3D6811CA Synthonix Corporation

6-Nitroheptan-3-one

  • MDL number No MDL found
  • Cas Number : 83188-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43854.html

Safety Information

MSDS https://synthonix.com/43854.html
Availability details for
Quantity
  • Availability
  • Add to Cart