SY3H3D6811D2 Synthonix Corporation

6-Nitrobenzo[d]isoxazole

  • MDL number No MDL found
  • Cas Number : 39835-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44365.html

Safety Information

MSDS https://synthonix.com/44365.html
Availability details for
Quantity
  • Availability
  • Add to Cart