SY3H3D6811D9 Synthonix Corporation

2-Nitro-6-(trifluoromethyl)phenol

  • Cas Number : 1548-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44482.html

Safety Information

MSDS https://synthonix.com/44482.html
Availability details for
Quantity
  • Availability
  • Add to Cart