SY3H3D6811DA Synthonix Corporation

1-Nitro-2-(trifluoromethoxy)benzene

  • Cas Number : 1644-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44483.html

Safety Information

MSDS https://synthonix.com/44483.html
Availability details for
Quantity
  • Availability
  • Add to Cart