SY3H3D6811DE Synthonix Corporation

2-Nitro-3-(trifluoromethyl)aniline

  • MDL number No MDL found
  • Cas Number : 386-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44811.html

Safety Information

MSDS https://synthonix.com/44811.html
Availability details for
Quantity
  • Availability
  • Add to Cart