SY3H3D6811E0 Synthonix Corporation

4-Nitro-2-(trifluoromethoxy)aniline

  • MDL number MFCD18399915
  • Cas Number : 1261753-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44817.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44817.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart