SY3H3D6811E2 Synthonix Corporation

4-Nitrobenzo[d]thiazol-2-amine

  • Cas Number : 6973-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44885.html

Safety Information

MSDS https://synthonix.com/44885.html
Availability details for
Quantity
  • Availability
  • Add to Cart